Merck
  • Plasma histamine in patients with chronic renal failure and nephrotic syndrome.
Plasma histamine in patients with chronic renal failure and nephrotic syndrome.
Journal of clinical pathology (1991-03-01)
D S Gill, V A Fonseca, M A Barradas, R Balliod, J F Moorhead, P Dandona
ABSTRACT
Plasma histamine concentrations were measured using a commercially available monoclonal antibody radioimmunoassay in 38 patients with nephrotic syndrome, end stage renal failure, those receiving haemodialysis, and those receiving continuous ambulatory peritoneal dialysis to determine whether histamine may mediate damage to glomerular capillaries and arterial endothelium. Plasma histamine concentrations were significantly increased in all four patient groups when compared with those of controls and were the highest in two patients with pruritus. Raised plasma histamine concentrations in such patients are consistent with the hypothesis that histamine may contribute to the damage to glomerular capillaries and to arterial endothelium. These effects may be relevant to the pathogenesis of glomerular disease and atherosclerosis. Histamine may also contribute to the pathogenesis of pruritus in patients with chronic renal failure.
Product Description
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
p-Cresol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
SAFC
L-Proline
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
p-Cresol, ≥99%, FG