Merck
  • Determination of N-acetylcysteine in human plasma by gas chromatography-mass spectrometry.
Determination of N-acetylcysteine in human plasma by gas chromatography-mass spectrometry.
Journal of chromatography (1991-01-02)
A Longo, M Di Toro, C Galimberti, A Carenzi
ABSTRACT
An automatic mass spectrometric method for the quantitation of N-acetylcysteine (NAC) in human plasma has been developed. NAC was extracted from plasma with ethyl acetate and derivatized in two steps with 2-propanol and pentafluoropropionic anhydride. The volatile derivative obtained was ideal for gas chromatographic-mass spectrometric analysis. Data obtained by analysing the plasma of healthy volunteers to whom 600 mg of NAC had been orally given are reported.
Product Description
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent