Merck
  • Detection of autosomal dominant polycystic kidney disease by NMR spectroscopic fingerprinting of urine.
Detection of autosomal dominant polycystic kidney disease by NMR spectroscopic fingerprinting of urine.
Kidney international (2011-03-11)
Wolfram Gronwald, Matthias S Klein, Raoul Zeltner, Bernd-Detlef Schulze, Stephan W Reinhold, Markus Deutschmann, Ann-Kathrin Immervoll, Carsten A Böger, Bernhard Banas, Kai-Uwe Eckardt, Peter J Oefner
ABSTRACT
Autosomal dominant polycystic kidney disease (ADPKD) is a frequent cause of kidney failure; however, urinary biomarkers for the disease are lacking. In a step towards identifying such markers, we used multidimensional-multinuclear nuclear magnetic resonance (NMR) spectroscopy with support vector machine-based classification and analyzed urine specimens of 54 patients with ADPKD and slightly reduced estimated glomerular filtration rates. Within this cohort, 35 received medication for arterial hypertension and 19 did not. The results were compared with NMR profiles of 46 healthy volunteers, 10 ADPKD patients on hemodialysis with residual renal function, 16 kidney transplant patients, and 52 type 2 diabetic patients with chronic kidney disease. Based on the average of 51 out of 701 NMR features, we could reliably discriminate ADPKD patients with moderately advanced disease from ADPKD patients with end-stage renal disease, patients with chronic kidney disease of other etiologies, and healthy probands with an accuracy of >80%. Of the 35 patients with ADPKD receiving medication for hypertension, most showed increased excretion of proteins and also methanol. In contrast, elevated urinary methanol was not found in any of the control and other patient groups. Thus, we found that NMR fingerprinting of urine differentiates ADPKD from several other kidney diseases and individuals with normal kidney function. The diagnostic and prognostic potential of these profiles requires further evaluation.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Hippuric acid, 98%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
β-Alanine, 99%
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG