Merck
Hydroquinone concentrations in skin lightening creams.
The British journal of dermatology (1986-04-01)
J Boyle, C T Kennedy
ABSTRACT
Forty-one over-the-counter skin lightening creams were analysed for hydroquinone content, and the accuracy of tables of contents supplied with these products was assessed. Eight of the 41 were found to contain more than 2% hydroquinone, the maximum concentration permitted by the U.K. Cosmetic Products Regulations. Eighteen of these preparations failed to comply with labelling requirements. A list is given of those products containing less than 2% hydroquinone which are adequately labelled.
Product Description
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications