Merck
  • Determination of ammonia and urea in urine and of urea in blood by use of an ammonia-selective electrode.
Determination of ammonia and urea in urine and of urea in blood by use of an ammonia-selective electrode.
Clinical chemistry (1979-11-01)
J Georges
PMID498498
ABSTRACT
I propose a single, quick method for measuring ammonia in urine and urea in plasma and urine. An ammonia-selective electrodie is used, set up on a microcell, which allows use of small sample volumes. Ammonia is measured directly after partial conversion of ammonium ions to NH3. Urea is measured after its hydrolysis by urease. With urine, the two procedures can be carried out successively in the same cell and on the same sample without changing the procedural conditions. Linear electrode response and accuracy have been checked for concentrations in the expected (normal) range.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Urea, European Pharmacopoeia (EP) Reference Standard