Merck
Radioimmunoassay of 2-hydroxyestrone.
Steroids (1978-02-01)
P Ball, G Emons, O Haupt, H O Hoppen, R Knuppen
PMID663967
ABSTRACT
Under the protection of ascorbic acid a 2-hydroxyestrone bovine serum albumin conjugate was prepared containing intact 2-hydroxyestrone as determined by gas chromatographymass spectometry. Using this antigen highely specific antibodies were raised in rabbits. Cross-reactivity for 2-hydroxyestradiol and 2-hydroxyestriol was 26 and 4.5%, respectively. An assay procedure of 2-hydroxyestrone in human plasma is described. Using special precautions the assay allows the determination of 2-hydroxyestrone in plasma samples of women (50-95 pg/ml), pregnant women (105-220 pg/ml), men (45-65 pg/ml) and children(20-40 pg/ml).
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Estriol 16α-(β-D-glucuronide), ≥97%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard