Merck
  • Excretion of amines and their metabolites by two patients in hepatic coma treated with L-dopa.
Excretion of amines and their metabolites by two patients in hepatic coma treated with L-dopa.
Clinical pharmacology and therapeutics (1983-09-01)
G M Tyce, J Stockard, N S Sharpless, M D Muenter
ABSTRACT
Two patients in hepatic coma were treated with L-dopa. The first patient showed clear clinical improvement, but the second patient did not. Analyses of urinary metabolites indicated that L-dopa was not absorbed by the second patient. There was evidence that L-dopa had the following beneficial effects in the first patient: (1) increased production of urine, which could have been accompanied by increased excretion of toxins; (2) displacement of tyramine from transmitter sites (because increased excretion of p-hydroxyphenylacetic acid, a major metabolite of tyramine, occurred during L-dopa treatment in patient 1); (3) replenishment of dopamine, and to a much lesser extent, norepinephrine, at central or peripheral neuroeffector junctions; and (4) scavenging of methyl groups by L-dopa, because ratio of methylated amines to catecholamines was higher than normal in both comatose patients before L-dopa treatment, and this ratio decreased during L-dopa treatment in patient 1.
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%