Merck
Quantification of endogenous aliphatic alcohols in serum and urine.
Journal of chromatography (1982-04-30)
H M Liebich, H J Buelow, R Kallmayer
ABSTRACT
The endogenous aliphatic alcohols ethanol, n-propanol, n-butanol, isobutanol and isopentanol in serum and urine were measured by gas chromatography-mass fragmentography. Whereas for the higher-molecular-weight alcohols extraction with dichloromethane is used, ethanol is determined by direct injection of serum and urine. When zero is assigned to all values below the detection limits of the procedures (0.1 mg/l for ethanol and 2 micrograms/l for each of the higher-molecular-weight alcohols) the following normal ranges are found: ethanol, 0-39 mg/l in serum and 0-46 mg per 24 h in urine; n-propanol, 0-48 micrograms/l in serum and 0-300 micrograms per 24 h in urine; n-butanol, 0-20 micrograms/l in serum and 0-18 micrograms per 24 h in urine. The isobutanol and isopentanol levels in the serum and urine of normal subjects are below the detection limit. For diabetic patients, on average increased levels are found for ethanol, n-propanol and n-butanol in serum and urine.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
3-Methyl-L-histidine
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Methylglyoxal solution, ~40% in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylglyoxal solution, technical, ~40% in H2O
Sigma-Aldrich
Dimethylamine solution, purum, 33% in absolute ethanol (~5.6 M)
Sigma-Aldrich
1-Methyl-L-histidine, ≥98.0% (TLC)
Sigma-Aldrich
(R)-3-Hydroxybutyric acid, ≥98.0% (T)
Sigma-Aldrich
4-Heptanone, 98%
Sigma-Aldrich
4-Heptanone, ≥97%, FG
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Heptanone, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ≥99%, FCC, FG