Merck
  • The association between measured and clinically assessed amniotic fluid volume and fetal growth and nutrition.
The association between measured and clinically assessed amniotic fluid volume and fetal growth and nutrition.
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde (1978-09-16)
K D Gunston, D D Davey
PMID734583
ABSTRACT
A close correlation was found between amniotic fluid volume measured by using para-aminohippuric acid and the clinical assessment of volume. No correlation was found between measured amniotic fluid volume and fetal weight, length, or weight for gestational age.
Sigma-Aldrich
p-Aminohippuric acid, ≥99%