Merck
  • Higher levels of plasma cholesterol sulfate in patients with liver cirrhosis and hypercholesterolemia.
Higher levels of plasma cholesterol sulfate in patients with liver cirrhosis and hypercholesterolemia.
Lipids (1993-09-01)
N Tamasawa, A Tamasawa, K Takebe
ABSTRACT
An analytical method for the determination of cholesterol sulfate (CS) in plasma using gas-liquid chromatography was developed. We measured plasma CS concentrations in patients with liver cirrhosis and hypercholesterolemia as examples of disorders that involve aberrations in cholesterol metabolism. Patients with liver cirrhosis had plasma CS concentrations that were significantly higher than those of control subjects (444.6 +/- 51.7 vs. 253.0 +/- 24.6 micrograms/dL, mean +/- SE). The levels of other lipids were lower in cirrhotics, although the differences were not significant. There was no correlation between the levels of CS and sulfated bile acids in cirrhotic patients. CS levels in plasma were also higher in subjects with hypercholesterolemia (413.7 +/- 44.5 micrograms/dL); however, the ratio of CS to total cholesterol (TC) clearly differed between cirrhotics and hypercholesterolemic subjects (1.44 +/- 0.11 x 10(-3) vs. 3.31 +/- 0.63 x 10(-3); P < 0.05). Both in subjects with hypercholesterolemia and in healthy controls, the CS/TC ratio was similar and CS accounted for roughly 0.14% of the TC concentration.
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
Cholesteryl palmitate, ≥98% (HPLC; detection at 205 nm)
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Aspartic acid
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG