Merck
  • Determination of tryptophan and its kynurenine pathway metabolites in human serum by high-performance liquid chromatography with simultaneous ultraviolet and fluorimetric detection.
Determination of tryptophan and its kynurenine pathway metabolites in human serum by high-performance liquid chromatography with simultaneous ultraviolet and fluorimetric detection.
Journal of chromatography. B, Biomedical applications (1996-01-12)
C Hervé, P Beyne, H Jamault, E Delacoux
ABSTRACT
An isocratic reversed-phase high-performance liquid chromatographic method for the simultaneous determination of tryptophan and four metabolites of the kynurenine pathway (kynurenine, 3-hydroxykynurenine, kynurenic acid and 3-hydroxyanthranilic acid) in human serum is described. This new method, which uses both isocratic elution and two on-line connected programmable ultraviolet and spectrofluorimetric detectors, allows the determination of these metabolites, in the physiological ranges, with satisfying specificity and sensitivity within 30 min.
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)
Sigma-Aldrich
3-Hydroxyanthranilic acid, 97%