Merck
  • The Synthesis of Substituted Penicillins and Simpler Structural Analogs. III. Phthalimido ?-Lactam-Thiazolidines Derived from Penicillamine.
The Synthesis of Substituted Penicillins and Simpler Structural Analogs. III. Phthalimido ?-Lactam-Thiazolidines Derived from Penicillamine.
Journal of the American Chemical Society (1951)
Sheehan JC, et al.