Merck
  • Characterisation of the ester-substituted products of the reaction of p-t-butyl calix[4]arene and ethyl bromoacetate using LC-UV-MS and LC-DAD.
Characterisation of the ester-substituted products of the reaction of p-t-butyl calix[4]arene and ethyl bromoacetate using LC-UV-MS and LC-DAD.
Talanta (2008-10-31)
Gillian McMahon, Rachel Wall, Kieran Nolan, Dermot Diamond
ABSTRACT
A series of derivatisation reactions between p-t-butyl calix[4]arene and ethyl bromoacetate were carried out in order to prepare 1,3 diester substituted calix[4]arene. Mass spectral data, obtained from direct injection of samples, indicated that the reactions were rich in the desired product. Since the ultra violet (UV) spectra of the desired product and possible impurities are very similar, liquid chromatography (LC) chromatographic data seemed to corroborate these results. However, when on-line LC-UV-MS was carried out and each LC peak subjected to MS analysis as it eluted, a very different picture emerged. It was found that many of these reactions actually contained high levels of the monoester product which, having less affinity for sodium in the MS, is therefore seriously underestimated in any direct injection assay. LC-diode array detection (DAD) methods were also used to help successfully identify and characterise the compounds being formed in these complex reactions. The overall results obtained in this paper allowed the optimal reaction conditions to be determined for this reaction. LC-MS analysis of the chromatographic peaks also identified the presence of two isomers of the diester substituted calix[4]arene (1,3 and 1,2 diesters). The combination of LC and UV/MS detection is required for accurate analysis of the products of such reactions.
Sigma-Aldrich
Ethyl bromoacetate, reagent grade, 98%
Pricing and availability is not currently available.