Merck
Characterization of adhesion molecules on human myeloma cell lines.
Blood (1992-11-01)
H Uchiyama, B A Barut, D Chauhan, S A Cannistra, K C Anderson
ABSTRACT
In multiple myeloma, malignant plasma cells are localized in marrow and rarely circulate in peripheral blood. To investigate the role of adhesion proteins in this process, we determined the expression and function of adhesion molecules on cell lines derived from patients with myeloma. The U266, ARH-77, IM-9, and HS-Sultan cell lines strongly expressed beta 1 and alpha 4 integrins (89% to 98% positive), confirming that VLA-4 is the principal integrin on these cell lines. The U266 and IM-9 cell lines also expressed alpha 3 integrin on 15% to 20% cells. In contrast, all lines lacked cell surface alpha 2, alpha 5, and alpha 6 integrin expression (< 5% positive). These cell lines adhered to fibronectin (20% to 40% specific binding), without significant binding to either collagen or laminin. Adhesion of these cell lines to fibronectin was partially blocked with either anti-beta 1 integrin monoclonal antibody (MoAb) (75% inhibition), anti-alpha 4 integrin MoAb (75% inhibition), or RGD peptide (50% inhibition), but was unaffected by anti-alpha v beta 3 or anti-alpha IIb beta 3 MoAbs. Moreover, the combination of anti-beta 1 plus RGD peptide or anti-alpha 4 plus RGD peptide inhibited binding to fibronectin by 80% and 95%, respectively. Finally, pretreatment and coculture of the IM-9 cell line with interleukin-6 (IL-6) resulted in a 52% decrease in specific binding to fibronectin (30% +/- 6% to 15% +/- 6%; P = .001), associated with a decrease in the number of cells expressing VLA-4 and a decrease in intensity of VLA-4 expression. These data suggest that myeloma cells adhere to fibronectin through VLA-4 as well as through RGD-dependent mechanisms, and that this binding can be downregulated by IL-6. Future studies of binding of both myeloma cell lines and freshly isolated tumor cells to extracellular matrix proteins and to marrow stroma may enhance our understanding of localization and trafficking of cells within the bone marrow microenvironment.
Product Description
IM 9, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86051302, human bone marrow, Lymphoblast