Merck
  • Pig liver esterase. Reactions with alcohols, structure-reactivity correlations, and the acyl-enzyme intermediate.
Pig liver esterase. Reactions with alcohols, structure-reactivity correlations, and the acyl-enzyme intermediate.
Biochemistry (1971-03-30)
P Greenzaid, W P Jencks
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid