Merck
  • Rapid and sensitive galactose oxidase-peroxidase biosensor for galactose detection with prolonged stability
Rapid and sensitive galactose oxidase-peroxidase biosensor for galactose detection with prolonged stability
Biotechnol. Tech. (1999)
Tkac J, et al.