Merck
Clinical evaluation of C-Film, a vaginal contraceptive.
The Journal of international medical research (1975-01-01)
O Frankman, N Raabe, C A Ingemansson
Product Description
Silver, foil, not light tested, 25x25mm, thickness 0.01mm, 99.95+%
Silver, foil, not light tested, 25x25mm, thickness 0.004mm, 99.97%
Silver, foil, not light tested, 50x50mm, thickness 0.008mm, 99.97%
Silver, foil, not light tested, 25x25mm, thickness 0.025mm, annealed, 99.95+%
Silver, foil, not light tested, 150x150mm, thickness 0.008mm, 99.97%
Silver, foil, not light tested, 100x100mm, thickness 0.008mm, 99.97%
Silver, foil, not light tested, 25x25mm, thickness 0.001mm, 99.97%
Silver, foil, not light tested, 25x25mm, thickness 0.008mm, 99.97%
Silver, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 0.02mm, 99.97%
Silver, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95+%
Silver, foil, 25mm disks, thickness 0.008mm, 99.97%
Silver, foil, 6mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Silver, foil, light tested, 100x100mm, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Silver, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.95+%
Silver, foil, 8mm disks, thickness 0.008mm, 99.97%
Silver, foil, 8mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 8mm disks, thickness 0.015mm, 99.97%
Silver, foil, 8mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 8mm disks, thickness 0.003mm, 99.9%
Silver, foil, 6mm disks, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 6mm disks, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%