- Lead foil technique for partially edentulous radiographic guide.
Lead foil technique for partially edentulous radiographic guide.
The Journal of prosthetic dentistry (2012-10-04)
Jason Chong, Wook-Jin Seong, Heather J Conrad
PMID23031734
Product Description
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.008mm, 99.99+%
Lead, foil, light tested, 100x100mm, thickness 0.025mm, 99.99+%
Lead, foil, 25x25mm, thickness 3.0mm, as rolled, 99.99+%
Lead, foil, 50mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 8mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 6mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 50mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%