Product Description
Tungsten, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.6mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.016mm, coil width .36mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.035mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.05mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.025mm, coil width .7mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.6mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.035mm, coil width .5mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.15mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.3mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.6mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.08mm, coil width .9mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.10mm, coil width .9mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.035mm, coil width .75mm, as rolled, 99.95%