Merck
  • 1H NMR relaxivity of aqueous suspensions of titanium dioxide nanoparticles coated with a gadolinium (III) chelate of a DOTA-monoamide with a phenylphosphonate pendant arm.
<SUP>1</SUP>H NMR relaxivity of aqueous suspensions of titanium dioxide nanoparticles coated with a gadolinium (III) chelate of a DOTA-monoamide with a phenylphosphonate pendant arm.
Journal of Materials Chemistry (2009)
Rehor I, et al.