Merck
  • An Improved Synthesis of Phenyl (trichloro-methyl) mercury from Sodium Methoxide and Ethyl Trichloroacetate.
An Improved Synthesis of Phenyl (trichloro-methyl) mercury from Sodium Methoxide and Ethyl Trichloroacetate.
The Journal of Organic Chemistry (1963)
Schweizer EE and O'Neill GJ.