Merck
  • A sensitive gas chromatographic/mass spectrometric method for the resolution and quantification of ethosuximide enantiomers in biological fluids.
A sensitive gas chromatographic/mass spectrometric method for the resolution and quantification of ethosuximide enantiomers in biological fluids.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2002-05-15)
Lino Sghendo, Janet Mifsud, Roger Ellul-Micallef, Joe Portelli, Jeff S Millership
ABSTRACT
A modified specific, sensitive and reproducible chiral gas chromatographic (GC) method for the resolution and quantification of ethosuximide enantiomers in urine and plasma was developed. The samples were extracted by liquid-liquid extraction, using diethylether and the enantiomers were separated and quantified on a chiral gas chromatographic column (25QC2 / CYDEX- beta 0.25). The method involved the use of GC/MS instrumentation for the acquisition of data in the electron impact selective-ion monitoring mode, collecting ions characteristic of both ethosuximide and alpha, alpha - dimethyl - beta - methylsuccinimide, the internal standard and of mass-to-charge ratio (m/z) exactly equal to 55 and 70 units. The limit of quantitation of the method was 2.5 microg/ml for both urine and plasma with both enantiomers. The method proved to be linear, precise and reproducible in the 5-300 microg/ml concentration range for urine samples and in the 10-250 microg/ml concentration range for plasma samples. Future research work envisaged the application of this method in pharmacokinetic and pharmacodynamic studies.