Merck
  • Tribromo melamine as novel and versatile catalyst for the formylation and acetylation of alcohols.
Tribromo melamine as novel and versatile catalyst for the formylation and acetylation of alcohols.
Chinese Journal of Catalysis (2013)
Hajjami M, et al.