Merck
  • Selective chemoprecipitation to enrich nitropeptides from complex proteomes for mass-spectrometric analysis.
Selective chemoprecipitation to enrich nitropeptides from complex proteomes for mass-spectrometric analysis.
Nature protocols (2014-03-22)
Laszlo Prokai, Jia Guo, Katalin Prokai-Tatrai
ABSTRACT
Post-translational protein nitration has attracted interest owing to its involvement in cellular signaling, effects on protein function and potential as biomarker of nitroxidative stress. We describe a procedure for enriching nitropeptides for mass spectrometry (MS)-based proteomics that is a simple and reliable alternative to immunoaffinity-based methods. The starting material for this procedure is a proteolytic digest. The peptides are reacted with formaldehyde and sodium cyanoborohydride to dimethylate all the N-terminal and side chain amino groups. Sodium dithionite is added subsequently to reduce the nitro groups to amines; in theory, the only amino groups present will have originally been nitro groups. The peptide sample is then applied to a solid-phase active ester reagent (SPAER), and those peptides with amino groups will be selectively and covalently captured. Release of the peptides on hydrolysis with trifluoroacetic acid (TFA) results in peptides that have a 4-formyl-benzamido group where the nitro group used to be. In qualitative setups, the procedure can be used to identify proteins modified by reactive nitrogen species and to determine the specific sites of their nitration. Quantitative measurements can be performed by stable-isotope labeling of the peptides in the reductive dimethylation step. Preparation of the SPAER takes about 1 d. Enrichment of nitropeptides requires about 2 d, and sample preparations need 1-30 h, depending on the experimental design. LC-MS/MS assays take from 4 h to several days and data processing can be done in 1-7 d.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1
Sigma-Aldrich
Universal Proteomics Standard Set, Protein Mass Spectrometry Calibration Standard
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%