Merck
  • A naturalistic evaluation and audit database of agomelatine: clinical outcome at 12 weeks.
A naturalistic evaluation and audit database of agomelatine: clinical outcome at 12 weeks.
Acta psychiatrica Scandinavica (2012-12-18)
A Sparshatt, R H McAllister Williams, D S Baldwin, P M Haddad, S Bazire, E Weston, P Taylor, D Taylor
ABSTRACT
To determine the effectiveness of agomelatine in routine clinical practice and explore factors associated with response and continuation. Consecutive patients prescribed agomelatine in participating psychiatric services were included. Patient demographic and outcome data were collected at treatment initiation and then at weeks 4, 8 and 12. Outcomes were analysed with respect to clinical and demographic factors. A total of 110 patients from nine NHS trusts were followed through 12 weeks of treatment. Agomelatine was largely used in difficult-to-treat or refractory patients: 83 (75%) had failed to respond to, or relapsed on, prior antidepressants. There were high rates of physical (54.5%) and psychiatric (50.0%) comorbidity. At 12 weeks of treatment, 68 (62%) continued agomelatine treatment. Overall, 69 subjects (62.7%) improved by at least one point of the Clinical Global Impression (severity) scale. Of 42 who discontinued, 23 (56%) discontinued because of lack of efficacy and 10 (24%) due to an adverse event. Of all variables examined, only a history of more than five episodes of depression significantly predicted discontinuation of treatment (OR continuation - 0.36, 95% CI 0.14, 0.95). Agomelatine was effective and generally well tolerated in a cohort of difficult-to-treat patients in clinical practice.
Sigma-Aldrich
Agomelatine, ≥98% (HPLC)