Merck
  • Efficacy of bath and orally administered praziquantel and fenbendazole against Lepidotrema bidyana Murray, a monogenean parasite of silver perch, Bidyanus bidyanus (Mitchell).
Efficacy of bath and orally administered praziquantel and fenbendazole against Lepidotrema bidyana Murray, a monogenean parasite of silver perch, Bidyanus bidyanus (Mitchell).
Journal of fish diseases (2013-03-16)
J M Forwood, J O Harris, M R Deveney
ABSTRACT
We investigated the efficacy of praziquantel (PZQ) and fenbendazole (FBZ), each administered by bath and orally, against the monogenean Lepidotrema bidyana Murray, a gill parasite of the freshwater fish silver perch, Bidyanus bidyanus (Mitchell). PZQ and FBZ were each administered by bath at 10 mg L⁻¹ for 48 h and on surface-coated feed pellets at 75 mg kg⁻¹ per body weight (BW) per day for 6 days. Bath treatments of PZQ and FBZ had an efficacy of 99% and 91%, respectively, against adult L. bidyana. Oral treatments of PZQ and FBZ had an efficacy of 79% and 95%, respectively, against adult L. bidyana. Fish rejected feed pellets surface-coated with PZQ, suggesting that palatability of surface-coated PZQ-medicated feed is poor, which undermined efficacy. In all trials, some juvenile parasites were present on fish after treatment during efficacy assessment, indicating that efficacy may be lower against juvenile parasites or that recruitment occurred post-treatment, demonstrating that repeat treatments are necessary to effectively control L. bidyana in aquaculture.
Sigma-Aldrich
Fenbendazole, ≥98%
Supelco
Fenbendazole, VETRANAL®, analytical standard
Supelco
Praziquantel, VETRANAL®, analytical standard