Merck
  • Enhanced transformation and dechlorination of p-chloronitrobenzene in the combined ZVI-anaerobic sludge system.
Enhanced transformation and dechlorination of p-chloronitrobenzene in the combined ZVI-anaerobic sludge system.
Environmental science and pollution research international (2013-03-29)
Liang Zhu, Haizhuan Lin, Jiaoqin Qi, Xiangyang Xu
ABSTRACT
The combination of zero-valent iron (ZVI) with anaerobic sludge for enhancing reductive transformation and dechlorination of p-chloronitrobenzene (p-ClNB) was investigated in this study. p-ClNB was quickly reduced into p-chloroaniline (p-ClAn) and subsequently dechlorinated into aniline in the complex system, and the strengthening factor for pseudo-first-order transformation rate constant of p-ClNB (Q, k ZVI + sludge/(k sludge + k ZVI)) was above 3. The Q values for the different ZVI types with anaerobic sludge were as following: Reduced ZVI (RZVI) > Industrial ZVI > Nanoscale ZVI (NZVI). Thereinto, the aggregation of NZVI occurred, and its reaction activity declined. Furthermore, the increase of ZVI dosage promoted the p-ClNB transformation, but the p-ClAn dechlorination rate and Q value were not improved. With the anaerobic biomass increasing, the dechlorination rate of p-ClAn was significantly enhanced, and the Q value had positive relation with the mass ratio of anaerobic sludge to RZVI.
Sigma-Aldrich
1-Chloro-4-nitrobenzene, 99%