Merck
  • Polypyrrole-poly(3,4-ethylenedioxythiophene)-Ag (PPy-PEDOT-Ag) nanocomposite films for label-free electrochemical DNA sensing.
Polypyrrole-poly(3,4-ethylenedioxythiophene)-Ag (PPy-PEDOT-Ag) nanocomposite films for label-free electrochemical DNA sensing.
Biosensors & bioelectronics (2013-04-13)
S Radhakrishnan, C Sumathi, Ahmad Umar, Sang Jae Kim, J Wilson, V Dharuman
ABSTRACT
The electrochemical DNA hybridization sensing of bipolymer polypyrrole and poly(3,4-ethylenedioxythiophene) (PPy-PEDOT) nanotubes functionalized with Ag nanoparticles has been investigated. The bipolymer nanotubes are prepared by simple chemical route and silver nanoparticles (Ag) further deposited over the PPy-PEDOT nanotubes to form PPy-PEDOT-Ag nanocomposite films. DNA labeled at 5'end using 6-mercapto-1-hexhane (HS-ssDNA) is immobilized on the PPy-PEDOT-Ag surface to form PPy-PEDOT-Ag-S-ssDNA and hybridization sensing is done in phosphate buffer. The presence of Ag nanoparticles (~28±5nm) well dispersed in the polymer composite with high surface area, high electrical conductivity and catalytic activity provides desirable microenvironment for the immobilization of probe DNA with controlled orientation leading to increased hybridization efficiency with target DNA. The morphological and structural characterizations by a scanning electron microscope (SEM) and X-ray diffraction (XRD) confirm the nanotube structure of composite polymer while Raman measurements indicate the efficient interactions between the PPy, PEDOT, Ag and HS-ssDNA. The sensor effectively discriminates different target DNA sequences with PPy-PEDOT-Ag-S-ssDNA substrate. The observed dynamic detection range is found between 1×10(-11)M and 1×10(-14)M with the lowest detection limit (3 σ/b) of 5.4×10(-15)M. This observed value is of higher sensitivity than that for MWCNT-Ag, PANi-Au, MWCNT-PPy-Au and PPy-PANi-Au composites reported previously.
Sigma-Aldrich
Polypyrrole, conductivity 10-50 S/cm (pressed pellet)
Sigma-Aldrich
Polypyrrole, doped, conductivity 0.5-1.5 S/cm (pressed pellet, typical), extent of labeling: ~5 wt. % loading, coated on titanium dioxide
Sigma-Aldrich
Polypyrrole, doped, conductivity 30 S/cm (bulk), extent of labeling: 20 wt. % loading, composite with carbon black