Merck
  • Application of permeability-limited physiologically-based pharmacokinetic models: part II - prediction of P-glycoprotein mediated drug-drug interactions with digoxin.
Application of permeability-limited physiologically-based pharmacokinetic models: part II - prediction of P-glycoprotein mediated drug-drug interactions with digoxin.
Journal of pharmaceutical sciences (2013-05-21)
Sibylle Neuhoff, Karen Rowland Yeo, Zoe Barter, Masoud Jamei, David B Turner, Amin Rostami-Hodjegan
ABSTRACT
Digoxin is the recommended substrate for assessment of P-glycoprotein (P-gp)-mediated drug-drug interactions (DDIs) in vivo. The overall aim of our study was to investigate the inhibitory potential of both verapamil and norverapamil on the P-gp-mediated efflux of digoxin in both gut and liver. Therefore, a physiologically-based pharmacokinetic (PBPK) model for verapamil and its primary metabolite was developed and validated through the recovery of observed clinical plasma concentration data for both moieties and the reported interaction with midazolam, albeit a cytochrome P450 3A4-mediated DDI. The validated inhibitor model was then used in conjunction with the model developed previously for digoxin. The range of values obtained for the 10 trials indicated that increases in area under the plasma concentration-time curve (AUC) profiles and maximum plasma concentration observed (Cmax ) values of digoxin following administration of verapamil were more comparable with in vivo observations, when P-gp inhibition by the metabolite, norverapamil, was considered as well. The predicted decrease in AUC and Cmax values of digoxin following administration of rifampicin because of P-gp induction was 1.57- (range: 1.42-1.77) and 1.62-fold (range: 1.53-1.70), which were reasonably consistent with observed values of 1.4- and 2.2-fold, respectively. This study demonstrates the application of permeability-limited models of absorption and distribution within a PBPK framework together with relevant in vitro data on transporters to assess the clinical impact of modulated P-gp-mediated efflux by drugs in development.
Product Description
Supelco
Digoxin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Digoxin for peak identification, European Pharmacopoeia (EP) Reference Standard
Digoxin, European Pharmacopoeia (EP) Reference Standard
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-Norverapamil hydrochloride, ≥98% (HPLC)
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Supelco
Digoxin, analytical standard