Merck
  • Stability of polydopamine and poly(DOPA) melanin-like films on the surface of polymer membranes under strongly acidic and alkaline conditions.
Stability of polydopamine and poly(DOPA) melanin-like films on the surface of polymer membranes under strongly acidic and alkaline conditions.
Colloids and surfaces. B, Biointerfaces (2013-05-28)
Houliang Wei, Jun Ren, Bo Han, Li Xu, Lulu Han, Lingyun Jia
ABSTRACT
This study investigated the stability of polydopamine and poly(3,4-dihydroxyphenylalanine) (poly(DOPA)) melanin-like films on the surface of polymer substrates. Three polymer membranes, polypropylene (PP), poly(vinylidenefluoride) (PVDF) and nylon, were modified with polydopamine or poly(DOPA), and then immersed in 0.1M HCl or NaOH, followed by UV-vis spectrometry analysis to detect the presence of film detachment. The results showed that the outer parts of both polydopamine and poly(DOPA) films were detached, probably due to electrostatic repulsion between the polymers within the film, when the modified membranes were washed in HCl or NaOH solution. These two films were more stable in strongly acidic solution, but the stability of poly(DOPA) film was better than that of polydopamine film. Compared to the films on the surface of PVDF or nylon membrane, films on PP surface showed the lowest stability, possibly because of the hydrophobic property of PP. The process of film detachment was analyzed by GPC, which showed that unreacted dopamine or DOPA monomers were still present in the freshly formed films. The unreacted monomers, as well as polydopamine or poly(DOPA) that were incorporated in the film via noncovalent interactions, became detached when the film was exposed to strongly acidic or alkaline solution. Oxidation of freshly formed films could significantly enhance their stability. The results therefore provide us with a better understanding of the stability of melanin-like films, and allow us to develop an effective strategy for constructing stable films.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
3,4-Dihydroxy-DL-phenylalanine
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid