Merck
  • Strategies based on silica monoliths for removing pollutants from wastewater effluents: a review.
Strategies based on silica monoliths for removing pollutants from wastewater effluents: a review.
The Science of the total environment (2013-05-30)
Dina Rodrigues, Teresa A P Rocha-Santos, Ana C Freitas, Ana M P Gomes, Armando C Duarte
ABSTRACT
Silica monoliths have been used for more than half a century in a wide variety of applications, such as stationary phases for microextraction fibers, capillary columns for chromatography, in the encapsulation of biomolecules for affinity chromatography and for microfluidic or microarray chips and, more recently, and less well known for wastewater treatment. The main objective of this review article is to specifically overview the strategies that use silica monoliths for the removal of chemical pollutants from wastewater effluents or prepared solutions. The discussion of advantages and drawbacks of such strategies will be supported with the main studies carried out so far which have been performed in laboratory environment only. The application and potential research interest in several strategies using composites and biocomposites based silica monoliths as cleaning systems are also discussed.
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginic acid sodium salt, powder