Merck
  • In vitro biostability of poly(dimethyl siloxane/hexamethylene oxide)-based polyurethane/layered silicate nanocomposites.
In vitro biostability of poly(dimethyl siloxane/hexamethylene oxide)-based polyurethane/layered silicate nanocomposites.
Acta biomaterialia (2013-06-04)
Yosephine Andriani, Isabel C Morrow, Elena Taran, Grant A Edwards, Tara L Schiller, Azlin F Osman, Darren J Martin
ABSTRACT
We have prepared a number of silicone-based thermoplastic polyurethane (TPU) nanocomposites and demonstrated an enhancement of in vitro biostability against metal-ion-induced oxidation for potential use in long-term implantable medical devices. Organoclays based on both low-aspect-ratio hectorites and high-aspect-ratio fluoromicas were evaluated after being dual-modified with two quaternary alkyl ammonium salts with differing degrees of polarity. The resultant nanocomposites were tested for in vitro biostability using physiologically relevant oxidizing conditions. Subsequently, the effects of oxidative treatment on the surface degradation and bulk mechanical integrity of the nanocomposites were investigated and compared with the parent TPUs to identify nanocomposites with the most desirable features for long-term implantation. Here, we demonstrate that the low-aspect-ratio organohectorite was delaminated and well dispersed in the nanocomposites. Importantly, these factors gave rise to the enhanced oxidative stability. In addition, the mechanical properties of all nanocomposites were less adversely affected by the oxidative treatment compared to their parent TPUs. These results suggest the potential for improved mechanical integrity and biostability when suitable dual modified organoclays are incorporated in a silicone-based TPU.
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)