Merck
  • Electrochemical detection of toxic ractopamine and salbutamol in pig meat and human urine samples by using poly taurine/zirconia nanoparticles modified electrodes.
Electrochemical detection of toxic ractopamine and salbutamol in pig meat and human urine samples by using poly taurine/zirconia nanoparticles modified electrodes.
Colloids and surfaces. B, Biointerfaces (2013-06-05)
Muniyandi Rajkumar, Ying-Sheng Li, Shen-Ming Chen
ABSTRACT
Detection of ractopamine and salbutamol has been developed by employing the facile synthesis of poly taurine/zirconia nanoparticles (ZrO2) modified film glassy carbon electrode. The poly taurine/ZrO2 nanoparticles were directly utilized for the detection of ractopamine and salbutamol using linear sweep voltammetry (LSV). The modified electrode successfully shows the oxidation peak for ractopamine adsorption at 0.65V and salbutamol at 0.71V, which is purely based on the detection of adsorption signals of ractopamine and salbutamol, at the electrode surface. Furthermore, the electrochemical measurements and surface morphology were studied using cyclic voltammetry (CV), electrochemical impedance spectroscopy (EIS) and scanning electron microscopy (SEM) analysis. The modified electrode successfully detects the oxidation signals of ractopamine in the linear range of 1-28μM and salbutamol in the linear range of 5-220μM in laboratory samples. The proposed film also successfully detects the ractopamine signal (1-26μM) in pig meat samples and salbutamol signal (1-114μM) in human urine samples. It also exhibits two well-separated anodic oxidation peaks for uric acid and salbutamol in salbutamol-spiked human urine samples.
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
SAFC
Taurine
Sigma-Aldrich
Salbutamol
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Salbutamol hemisulfate salt, ≥98%