Merck
  • Subacute toxicity of copper and glyphosate and their interaction to earthworm (Eisenia fetida).
Subacute toxicity of copper and glyphosate and their interaction to earthworm (Eisenia fetida).
Environmental pollution (Barking, Essex : 1987) (2013-06-05)
Chui-Fan Zhou, Yu-Jun Wang, Cheng-Cheng Li, Rui-Juan Sun, Yuan-Chun Yu, Dong-Mei Zhou
ABSTRACT
Glyphosate (GPS) and copper (Cu) are common pollutants in soils, and commonly co-exist. Due to the chemical structure of GPS, it can form complexes of heavy metals and interface their bioavailability in soil environment. In order to explore the interactions between GPS and Cu, subacute toxicity tests of Cu and GPS on soil invertebrate earthworms (Eisenia fetida) were conducted. The relative weight loss and whole-worm metal burdens increased significantly with the increasing exposure concentration of Cu, while the toxicity of GPS was insignificant. The joint toxicity data showed that the relative weight loss and the uptake of Cu, as well as the superoxide dismutase, catalase and malondialdehyde activities, were significantly alleviated in the present of GPS, which indicated that GPS could reduce the toxicity and bioavailability of Cu in the soil because of its strong chelating effects.
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Pricing and availability is not currently available.
Supelco
Glyphosate, PESTANAL®, analytical standard
Pricing and availability is not currently available.