Merck
  • Application of green zero-valent iron nanoparticles to the remediation of soils contaminated with ibuprofen.
Application of green zero-valent iron nanoparticles to the remediation of soils contaminated with ibuprofen.
The Science of the total environment (2013-06-07)
S Machado, W Stawiński, P Slonina, A R Pinto, J P Grosso, H P A Nouws, J T Albergaria, C Delerue-Matos
ABSTRACT
Zero-valent iron nanoparticles (nZVIs) are often used in environmental remediation. Their high surface area that is associated with their high reactivity makes them an excellent agent capable of transforming/degrading contaminants in soils and waters. Due to the recent development of green methods for the production of nZVIs, the use of this material became even more attractive. However, the knowledge of its capacity to degrade distinct types of contaminants is still scarce. The present work describes the study of the application of green nZVIs to the remediation of soils contaminated with a common anti-inflammatory drug, ibuprofen. The main objectives of this work were to produce nZVIs using extracts of grape marc, black tea and vine leaves, to verify the degradation of ibuprofen in aqueous solutions by the nZVIs, to study the remediation process of a sandy soil contaminated with ibuprofen using the nZVIs, and to compare the experiments with other common chemical oxidants. The produced nZVIs had nanometric sizes and were able to degrade ibuprofen (54 to 66% of the initial amount) in aqueous solutions. Similar remediation efficiencies were obtained in sandy soils. In this case the remediation could be enhanced (achieving degradation efficiencies above 95%) through the complementation of the process with a catalyzed nZVI Fenton-like reaction. These results indicate that this remediation technology represents a good alternative to traditional and more aggressive technologies.
Product Description
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%