Merck
  • Enzymatic activity of cholesterol oxidase immobilized onto polymer nanoparticles mediated by Congo red.
Enzymatic activity of cholesterol oxidase immobilized onto polymer nanoparticles mediated by Congo red.
Colloids and surfaces. B, Biointerfaces (2013-06-12)
Rubens A Silva, Ana Maria Carmona-Ribeiro, Denise F S Petri
ABSTRACT
Poly(ethylene glycol), PEG, decorated polystyrene (PS) nanoparticles were synthesized and characterized by means of dynamic light scattering (DLS), zeta (ζ) potential measurements, Fourier transform infrared spectroscopy (FTIR) and scanning electron microscopy (SEM). The adsorption of Congo red (CR) onto PS/PEG particles was evidenced by the decrease of ζ potential values and increase in the particles mean diameter in comparison to bare particles. Cholesterol oxidase (ChOx), the main enzyme in the oxidation of cholesterol, adsorbed onto PS/PEG and PS/PEG/CR particles, as revealed by the increase in the particles mean size and spectrophotometry. The enzymatic activity of free and immobilized ChOx was determined as a function of time by means of a coupled reaction with horseradish peroxidase. The activity of free ChOx decreased with time, while the activity of immobilized ChOx increased with time; after 1h reaction the latter was half of the former. Freeze-drying the ChOx covered PS/PEG/CR particles allowed their storage for at least one month under room conditions without loss of enzymatic activity. Conjugation effects between CR and ChOx or cholesterol evidenced by circular dichroism and spectrophotometry rendered a conformational state of ChOx, such that the enzymatic action was favored. ChOx adsorbed onto PS/PEG presents no enzymatic activity, probably due to ChOx denaturation or unfavorable orientation. Freeze-dried and freshly prepared dispersions of ChOx immobilized onto PS/PEG/CR particles yielded linear response in the cholesterol concentration range of 100mgdL(-1) (lowest limit of normal blood concentration) to 300mgdL(-1) (high risk level).
Sigma-Aldrich
Cholesterol Oxidase from microorganisms, aqueous solution, ≥30 units/mg protein (biuret)
Sigma-Aldrich
Cholesterol Oxidase from microorganisms, lyophilized powder, ≥50 units/mg protein, recombinant, expressed in E. coli
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Congo Red, Dye content ≥35 %
Sigma-Aldrich
Cholesterol Oxidase from Streptomyces sp., lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
Cholesterol Oxidase microbial, recombinant, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Congo Red, certified by the BSC, BioXtra