Merck
  • Production of monosaccharides from napier grass by hydrothermal process with phosphoric acid.
Production of monosaccharides from napier grass by hydrothermal process with phosphoric acid.
Bioresource technology (2013-06-20)
Eri Takata, Ken Tsutsumi, Yuji Tsutsumi, Kenji Tabata
ABSTRACT
The production of monosaccharides from napier grass was investigated in the presence of acid catalysts using the hydrothermal process. When the napier grass was treated with 3 wt.% phosphoric acid at 160°C for 15min, the xylose yield reached 10.3 wt.%, corresponding to 72.0% of the xylan in it, whereas glucose was hardly obtained. A combined process was then conducted using an 85 wt.% phosphoric acid treatment at 60 °C for 1h followed by a hydrothermal treatment with 3 wt.% phosphoric acid. In the initial treatment with concentrated phosphoric acid the most of xylan was hydrolyzed to xylose, and the crystalline cellulose was converted to its amorphous form. The hydrolysis of cellulose to glucose was significantly enhanced during the following hydrothermal process with 3 wt.% phosphoric acid at 200 °C for 8 min. Consequently, 77.2% yield of xylose and 50.0% yield of glucose were obtained from the combined process.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O