Merck
  • Marine hydroid perisarc: a chitin- and melanin-reinforced composite with DOPA-iron(III) complexes.
Marine hydroid perisarc: a chitin- and melanin-reinforced composite with DOPA-iron(III) complexes.
Acta biomaterialia (2013-06-25)
Dong Soo Hwang, Admir Masic, Ekavianty Prajatelistia, Mihaela Iordachescu, J Herbert Waite
ABSTRACT
Many marine invertebrates utilize biomacromolecules as building blocks to form their load-bearing tissues. These polymeric tissues are appealing for their unusual physical and mechanical properties, including high hardness and stiffness, toughness and low density. Here, a marine hydroid perisarc of Aglaophenia latirostris was investigated to understand how nature designs a stiff, tough and lightweight sheathing structure. Chitin, protein and a melanin-like pigment, were found to represent 10, 17 and 60 wt.% of the perisarc, respectively. Interestingly, similar to the adhesive and coating of marine mussel byssus, a DOPA (3,4-dihydroxyphenylalanine) containing protein and iron were detected in the perisarc. Resonance Raman microprobe analysis of perisarc indicates the presence of catechol-iron(III) complexes in situ, but it remains to be determined whether the DOPA-iron(III) interaction plays a cohesive role in holding the protein, chitin and melanin networks together.
Sigma-Aldrich
3,4-Dihydroxy-DL-phenylalanine
Sigma-Aldrich
Chitin from shrimp shells, BioReagent, suitable for analysis of chitinase, purified powder
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder