Merck
  • Whey protein and alginate hydrogel microparticles for insulin intestinal absorption: evaluation of permeability enhancement properties on Caco-2 cells.
Whey protein and alginate hydrogel microparticles for insulin intestinal absorption: evaluation of permeability enhancement properties on Caco-2 cells.
International journal of pharmaceutics (2013-06-26)
Emmanuelle Déat-Lainé, Valérie Hoffart, Ghislain Garrait, Eric Beyssac
ABSTRACT
The evaluation of encapsulated insulin intestinal absorption enhancement was investigated by in vitro methods. Insulin-loaded microparticles (INS-MP) made of whey protein (WP) and alginate (ALG) were prepared by a cold gelation technique. Effect of INS encapsulation toward trypsin and chymotrypsin degradation was performed. Permeability studies using in vitro (Caco-2 cells) experiments were conducted. INS was partially protected by encapsulation toward enzymatic degradation. Moreover INS transport experiments showed that WP and, in lesser extent, ALG were able to enhance INS absorption both as MP and as polymeric solutions by opening the tight junctions. These experiments reinforced the interest of encapsulation in WP/ALG hydrogel combination.
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Alginic acid from brown algae, powder