Merck
  • Fermentation of biodiesel-derived glycerol by Bacillus amyloliquefaciens: effects of co-substrates on 2,3-butanediol production.
Fermentation of biodiesel-derived glycerol by Bacillus amyloliquefaciens: effects of co-substrates on 2,3-butanediol production.
Applied microbiology and biotechnology (2013-06-26)
Tao-Wei Yang, Zhi-Ming Rao, Xian Zhang, Mei-Juan Xu, Zheng-Hong Xu, Shang-Tian Yang
ABSTRACT
Cultivation in glycerol instead of sugars inhibits 2,3-butanediol (2,3-BD) production by Bacillus amyloliquefaciens. In this study, we report that B. amyloliquefaciens readily produces 2,3-BD from biodiesel-derived glycerol in the presence of beet molasses as a co-substrate. Unexpectedly, the molasses stimulated 2,3-BD production and simultaneously reduced the duration of fermentation. Productivity of 2,3-BD was enhanced at the start of fermentation, and yields increased under continuous molasses supply. Subsequently, 2,3-BD production in molasses-supplemented fed-batch culture was observed. Prior to inoculation of fed-batch fermentation culture, 15 g/l of molasses was added to the bioreactor. After 6 h of incubation, the bioreactor was fed with a solution containing 80 % glycerol and 15 % molasses. The 2,3-BD concentration, yield, and productivity significantly improved, reaching 83.3 g/l, 0.42 g/g, and 0.87 g/l·h, respectively. To our knowledge, these results are the highest report for 2,3-BD fermentation from biodiesel-derived glycerol.
Sigma-Aldrich
meso-2,3-Butanediol, 99%
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
(2S,3S)-(+)-2,3-Butanediol, 97%
Sigma-Aldrich
(2R,3R)-(−)-2,3-Butanediol, 97%