Merck
  • Diffuser-incorporated transmission NIR measurement for reliable analysis of packed granular samples.
Diffuser-incorporated transmission NIR measurement for reliable analysis of packed granular samples.
The Analyst (2013-06-28)
Jihye Yoon, Jaejin Kim, Pham Khac Duy, Mooeung Kim, Sukwon Kang, Hoeil Chung
ABSTRACT
A diffuser-incorporated transmission near-infrared (NIR) scheme that enables direct spectral collection of packed granular samples with reliable sample representation and reproducibility has been demonstrated. The analytical utility of this method has been evaluated for the determination of polyethylene (PE) pellet density and the discrimination of the geographical origin of rice samples. Based on the preliminary observation of transmission spectral features acquired from spherical polyoxymethylene (POM) packings composed of different particle sizes as well as packing thickness, a portion of the radiation was propagated through the void space in the packing without fully interacting with the POM pellets. This type of radiation, so-called non-fully interacted radiation (NFIR), adversely affected the sample representation as well as the reproducibility of transmission measurements. To maximize the interaction of NIR radiation with granular samples, a polytetrafluoroethylene (PTFE) diffuser was positioned in front of the sample packing to introduce isotropically diffused radiation into the sample. This diffuser-incorporated scheme resulted in highly reproducible transmission spectra for both packed granular samples. Consequently, the density determination of PE pellets as well as discrimination of rice samples according to geographical origin was more accurate using the proposed scheme.
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material