Merck
  • Plasma gel as a new augmentation material for injection laryngoplasty in rabbit model: histologic analysis.
Plasma gel as a new augmentation material for injection laryngoplasty in rabbit model: histologic analysis.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2013-07-13)
Seok Jin Hong, Sang Hyuk Lee, Sung Min Jin, Seon Uk Lee, Se Hyung Huh, Dong Hoon Kim, Soon Young Kwon, Seung Hoon Woo
ABSTRACT
Currently, a variety of materials are available for the treatment of glottal insufficiency. Ideal injection materials should be inexpensive, easily obtainable, nontoxic, and biocompatible. Plasma gel has recently been developed as an injectable, autologous material used in plastic surgery. The aim of this study was to evaluate the histological changes in rabbit vocal folds (VFs) after an injection of plasma gel, compared with collagen (Artecoll) or hyaluronic acid (Reviderm Intra). Experimental prospective animal study. Animal laboratory. Blood samples were collected from 12 New Zealand rabbits. Plasma collected from each rabbit was centrifuged and processed using a gel heating system. All rabbits received a 0.05 ml injection of plasma gel into the right VF, while an equivalent volume of Artecoll or Reviderm Intra was injected into the left VFs of each 6 rabbits randomly. The larynges were collected 2, 4, and 8 weeks after injection, and the tissues were stained for histological analysis. In comparison with left VFs injected with Artecoll or Reviderm Intra, there was significantly less inflammatory response and foreign body reaction in the plasma gel injected right VFs (P < .05). Multinucleated foreign body type giant cells were also more prevalent in the left VFs. There were no quantitative differences in the rates of neovascularization or collagen deposition between groups. Plasma gel is a biologically compatible material that may serve as a suitable augmentation material in injection laryngoplasty. Further studies that examine the long-term effects in a larger number of subjects are needed.
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)