Merck
  • A mitochondria-specific visible-light sensitized europium β-diketonate complex with red emission.
A mitochondria-specific visible-light sensitized europium β-diketonate complex with red emission.
Dalton transactions (Cambridge, England : 2003) (2013-07-16)
V Divya, Vandana Sankar, K G Raghu, M L P Reddy
ABSTRACT
Our recently developed Eu(3+) coordination compound, Eu(pfppd)3(tpy) [where Hpfppd = 4,4,5,5,5-pentafluoro-3-hydroxy-1-(phenanthren-3-yl)pentanedione and tpy = 2,2':6,6''-terpyridine] exhibits significant quantum yield (Φoverall = 41%) and long (5)D0 lifetime (880 μs) values under biologically relevant pH conditions (pH = 7.4) when excited with visible light. Hence, the Eu(3+) luminescent complex was examined for live cell imaging using the rat embryonic heart cell line, H9c2. The ternary Eu(3+) complex permeates into the H9c2 cells and co-localises with the mitochondria, as demonstrated by counterstaining experiments. Furthermore, the designed Eu(3+) bioprobe remains undissociated in the cell medium, showed a good cell permeability and a fast cellular uptake with a specific localization profile. Thus it has the potential to become a time-resolved imaging probe that is excitable in the visible-light range.
Product Description
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Sigma-Aldrich
Phenanthrene, 98%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
Phenanthrene, analytical standard, for environmental analysis