Merck
High-level secretory expression of metchnikowin in Escherichia coli.
Protein expression and purification (2013-07-23)
Di Wu, Yinghu Lu, Huoqing Huang, Lijuan Ma, Yuanyuan Che, Xiangdong Zha, Bin Yao, Peilong Yang
ABSTRACT
Metchnikowin is a proline-rich peptide from Drosophila with antibacterial and antifungal activities. Its commercial application is limited due to the low immune-inducible expression in vivo. Here we present a method to produce high-yield metchnikowin in recombinant Escherichia coli. The genes coding for metchnikowin and the fusion partner Cherry tag were subcloned into the pET22b vector to construct a recombinant expression plasmid and transformed into E. coli BL21 (DE3). The fusion protein was successfully expressed and secreted with a yield of 300μg/ml after 18-h induction. Active metchnikowin was released by cleavage of the Asp-Pro bond between fused proteins by 72-h formic acid hydrolysis at 50°C. After 24-h dialysis, metchnikowin was purified to electrophoretic homogeneity and showed significant antibacterial activities against both Bacillus subtilis and E. coli DH5α. It is the first report on efficient production of metchnikowin in recombinant E. coli.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Cesium formate, 98%
Supelco
Calcium formate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%