- Direct conversion of silver complexes to nanoscale hexagonal columns on a copper alloy for plasmonic applications.
Direct conversion of silver complexes to nanoscale hexagonal columns on a copper alloy for plasmonic applications.
Physical chemistry chemical physics : PCCP (2013-08-02)
Yuko S Yamamoto, Katsuyuki Hasegawa, Yuuki Hasegawa, Naoshi Takahashi, Yasutaka Kitahama, Satoshi Fukuoka, Norio Murase, Yoshinobu Baba, Yukihiro Ozaki, Tamitake Itoh
PMID23903026
ABSTRACT
We introduced a novel method for the rapid synthesis of silver nanohexagonal thin columns from an aqueous mixture of sodium thiosulfate (Na2S2O3) and silver chloride (AgCl) simply added to a phosphor bronze substrate. The reaction is based on galvanic displacement and the products are potentially useful for plasmonic applications.