Merck
  • Self-assembled films containing crude extract of avocado as a source of tyrosinase for monophenol detection.
Self-assembled films containing crude extract of avocado as a source of tyrosinase for monophenol detection.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Nirton C S Vieira, Reginaldo A Ferreira, Valquiria da Cruz Rodrigues, Francisco E G Guimarães, Alvaro A A de Queiroz
ABSTRACT
This paper reports on the use of the crude extract of avocado (CEA) fruit (Persea americana) as a source of tyrosinase enzyme. CEA was immobilized via layer by layer (LbL) technique onto indium tin oxide (ITO) substrates and applied in the detection of monophenol using a potentiometric biosensor. Poly(propylene imine) dendrimer of generation 3 (PPI-G3) was used as a counter ion in the layer by layer process due to its highly porous structure and functional groups suitable for enzyme linkage. After the immobilization of the crude CEA as multilayered films, standard samples of monophenol were detected in the 0.25-4.00 mM linear range with approximately 28 mV mM(-1) of sensitivity. This sensitivity is 14 times higher than the values found in the literature for a similar system. The results show that it is possible to obtain efficient and low-cost biosensors for monophenol detection using potentiometric transducers and alternative sources of enzymes without purification.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%