Merck
  • Green synthesis of silver nanoparticles using 4-acetamido-TEMPO-oxidized curdlan.
Green synthesis of silver nanoparticles using 4-acetamido-TEMPO-oxidized curdlan.
Carbohydrate polymers (2013-08-06)
Jing-Kun Yan, Pan-Fu Cai, Xiao-Qiang Cao, Hai-Le Ma, Qi Zhang, Nan-Zi Hu, Yi-Zhang Zhao
ABSTRACT
A facile, simple, and eco-friendly method using 4-acetamido-2,2,6,6-tetramethypiperidine-1-oxyl radical-oxidized curdlan (Oc) as both reducing and stabilizing agents was developed for the fabrication of silver nanoparticles (AgNPs) from silver nitrate (AgNO₃). The structure, morphology, and particle size of the as-prepared AgNPs were investigated by ultraviolet-visible spectroscopy, transmission electron microscopy, energy dispersive X-ray spectrometry, X-ray diffraction, and dynamic laser light scattering. The well-dispersed AgNPs were sphere like with a mean diameter of 15 nm. Their formation was dependent on reaction duration, reaction temperature, Oc concentration, and AgNO₃ concentration. Fourier transform-infrared and Raman spectra demonstrated that the as-prepared AgNPs can readily bind covalently with the carboxylate groups of Oc through the strong monodentate interaction in the reaction medium.
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
Sigma-Aldrich
Curdlan from Alcaligenes faecalis
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.