Merck
  • Reactivity in nucleophilic vinylic substitution (S(N)V):S(N)Vπ versus S(N)Vσ mechanistic dichotomy.
Reactivity in nucleophilic vinylic substitution (S(N)V):S(N)Vπ versus S(N)Vσ mechanistic dichotomy.
The Journal of organic chemistry (2013-08-07)
Israel Fernández, F Matthias Bickelhaupt, Einar Uggerud
ABSTRACT
The intrinsic electronic factors that determine reactivity in prototypical identity nucleophilic vinylic substitution reactions, X(-) + ViX → XVi + X(-) (Vi = vinyl), have been studied by performing quantum chemical calculations (OPBE/6-311++G(d,p)). Of the two limiting reaction types envisaged--the S(N)Vπ and S(N)Vσ mechanisms--the former is preferred for most combinations of nucleophiles and substrates, except for the combination of unactivated substrates and poor nucleophiles, as seen for the much studied reactions Cl(-) + CH2CHCl and Br(-) + CH2CHBr. It was found that periodic trends for S(N)Vπ are essentially the same as those previously reported for nucleophilic aromatic substitution, S(N)Ar, while intrinsic S(N)Vσ nucleophilicity parallels aliphatic S(N)2. It is therefore concluded that S(N)V reactivity in general can be understood in terms of this mechanistic dichotomy. Furthermore, a few representative reactions were analyzed applying two complementary schemes for energy decomposition analysis.
Product Description
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol