Merck
  • Nucleoside analogues with a 1,3-diene-Fe(CO)3 substructure: stereoselective synthesis, configurational assignment, and apoptosis-inducing activity.
Nucleoside analogues with a 1,3-diene-Fe(CO)3 substructure: stereoselective synthesis, configurational assignment, and apoptosis-inducing activity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-08-13)
Christoph Hirschhäuser, Juraj Velcicky, Daniel Schlawe, Erik Hessler, André Majdalani, Jörg-Martin Neudörfl, Aram Prokop, Thomas Wieder, Hans-Günther Schmalz
ABSTRACT
The synthesis and stereochemical assignment of two classes of iron-containing nucleoside analogues, both of which contain a butadiene-Fe(CO)3 substructure, is described. The first type of compounds are Fe(CO)3-complexed 3'-alkenyl-2',3'-dideoxy-2',3'-dehydro nucleosides (2,5-dihydrofuran derivatives), from which the second class of compounds is derived by formal replacement of the ring oxygen atom by a CH2 group (carbocyclic nucleoside analogues). These compounds were prepared in a stereoselective manner through the metal-assisted introduction of the nucleobase. Whilst the furanoid intermediates were prepared from carbohydrates (such as methyl-glucopyranoside), the carbocyclic compounds were obtained by using an intramolecular Pauson-Khand reaction. Stereochemical assignments based on NMR and CD spectroscopy were confirmed by X-ray structural analysis. Biological investigations revealed that several of the complexes exhibited pronounced apoptosis-inducing properties (through an unusual caspase 3-independent but ROS-dependent pathway). Furthermore, some structure-activity relationships were identified, also as a precondition for the design and synthesis of fluorescent and biotin-labeled conjugates.
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, meets USP testing specifications
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Biotin, ≥99.0% (T)
Biotin, European Pharmacopoeia (EP) Reference Standard