Merck
Four new lignans from Schisandra sphenanthera.
Journal of Asian natural products research (2013-08-16)
Yun-Fang Li, Yan Jiang, Jing-Fang Huang, Guang-Zhong Yang
ABSTRACT
Three new 7,8-secolignans, schisandlignans A-C (1, 2, and 4), one new dibenzocyclooctadiene lignan, schisandlignan D (5), together with nine known lignans 3',4'-dimethoxybenzoic acid (3″,4″-dimethoxyphenyl)-2-methyl-3-oxobutyl ester (3), gomisin J (6), rubrisandrin A(1b) (7), interiotherin B (8), schisantherin D (9), ( - )-machilusin (10), ganschisandrine (11), henricine A (12), and (+)-1-hydroxy pinoresinol (13), were isolated from the rattan of Schisandra sphenanthera. Their structures were determined by analysis of 1D and 2D NMR spectroscopic data.
Sigma-Aldrich
Pinoresinol, ≥95.0% (HPLC)
Supelco
Pinoresinol, analytical standard